missing translation for 'onlineSavingsMsg'
Learn More

CNGA3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18354791 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354791 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354791 Supplier Novus Biologicals Supplier No. NBP2847010.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNGA3 Polyclonal specifically detects CNGA3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGA3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias CCNC1, CCNCa, CCNCalpha, CNCG3, CNG channel alpha-3, CNG-3, CNG3ACHM2, cone photoreceptor cGMP-gated channel alpha subunit, Cone photoreceptor cGMP-gated channel subunit alpha, cyclic nucleotide gated channel alpha 3, cyclic nucleotide-gated cation channel alpha-3, Cyclic nucleotide-gated channel alpha-3
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human CNGA3
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1261
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.