missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNGA3 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the cyclic nucleotide-gated cation channel protein family which is required for normal vision and olfactory signal transduction. Mutations in this gene are associated with achromatopsia (rod monochromacy) and color blindness. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications
Specifications
| Antigen | CNGA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CNGA3 |
| Gene Accession No. | Q16281, Q9JJZ8 |
| Gene Alias | ACHM2; alpha subunit of cone photoreceptor CNG-channel; CCNC1; CCNCa; CCNCalpha; CNCG3; cng channel; CNG channel 1; CNG channel alpha-3; CNG1; CNG-1; CNG3; CNG-3; CNGA3; CNGalpha2; cone photoreceptor cGMP-gated channel alpha subunit; cone photoreceptor cGMP-gated channel subunit alpha; cyclic nucleotide gated channel alpha 3; cyclic nucleotide-gated cation channel alpha-3; Cyclic nucleotide-gated channel alpha-3; cyclic nucleotide-gated channel cone photoreceptor subunit alpha |
| Gene Symbols | CNGA3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?