missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNGA3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92423-0.02ml
This item is not returnable.
View return policy
Description
CNGA3 Polyclonal antibody specifically detects CNGA3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CNGA3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CCNC1, CCNCa, CCNCalpha, CNCG3, CNG channel alpha-3, CNG-3, CNG3ACHM2, cone photoreceptor cGMP-gated channel alpha subunit, Cone photoreceptor cGMP-gated channel subunit alpha, cyclic nucleotide gated channel alpha 3, cyclic nucleotide-gated cation channel alpha-3, Cyclic nucleotide-gated channel alpha-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human CNGA3 (NP_001289.1). MAKINTQYSHPSRTHLKVKTSDRDLNRAENGLSRAHSSSEETSSVLQPGIAMETRGLADSGQGSFTGQGIARLSRLIFLLRRWAARHVHHQDQGPDSFPDRFRGAELKEVSSQESNAQANVGSQEPADRGRSAWPLAKCNTNTSNNTEEEKKTKKKDAIVVDPSS | |
| 0.02 mL | |
| Neuroscience | |
| 1261 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction