missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNGA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553274
This item is not returnable.
View return policy
Description
Immunogen sequence: EAVTEYPDAK KVLEERGREI LMKEGLLDEN EVATSMEVDV QEKLGQLETN METLYTRFGR LLAEYTGAQQ KLKQRITVLE TKMKQNNEDD YLSDGMNSP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
CNGA2 represents the alpha subunit of a cyclic nucleotide-gated olfactory channel. The encoded protein contains a carboxy-terminal leucine zipper that mediates channel formation.
Specifications
| CNGA2 | |
| Polyclonal | |
| Unconjugated | |
| CNGA2 | |
| Cnca; CNCA1; CNCG2; Cncg4; cng channel; CNG channel 2; CNG channel alpha-2; CNG2; CNG-2; Cnga2; cyclic nucleotide gated channel 4; cyclic nucleotide gated channel alpha 2; cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha-2; cyclic nucleotide-gated olfactory channel; Cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2; OCNC1; OCNCa; OCNCALPHA; OLFCH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1260 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16280 | |
| CNGA2 | |
| Recombinant protein corresponding to Human CNGA2. Recombinant protein control fragment (Product #RP-91193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction