missing translation for 'onlineSavingsMsg'
Learn More

CNGA2 Antibody, Novus Biologicals™

Product Code. 18342863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342863 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342863 Supplier Novus Biologicals Supplier No. NBP305204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNGA2 Polyclonal specifically detects CNGA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGA2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:10-1:50, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3. with 0.05% Proclin 300
Gene Alias CNCA, CNCA1, CNCG2, CNG2CNG-2, cyclic nucleotide gated channel alpha 2, Cyclic nucleotide-gated cation channel 2, Cyclic nucleotide-gated channel alpha-2, cyclic nucleotide-gated olfactory channel, cyclic-nucleotide-gated cation channel 2, cyclic-nucleotide-gated olfactory channel, FLJ46312, OCNC1CNG channel alpha-2, OCNCa, OCNCalpha
Host Species Rabbit
Immunogen Recombinant protein of human CNGA2.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1260
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.