missing translation for 'onlineSavingsMsg'
Learn More

CNF1/CNF2 Antibody (JC4), Novus Biologicals™

Product Code. 18351772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351772 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351772 Supplier Novus Biologicals Supplier No. NBP311249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNF1/CNF2 Monoclonal specifically detects CNF1/CNF2 in E. coli samples. It is validated for Western Blot, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNF1/CNF2
Applications Western Blot, Functional Assay, Immunoassay
Classification Monoclonal
Clone JC4
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Cytotoxic necrotizing factor, Cytotoxic necrotizing factor type 1, Cytotoxic necrotizing factor type 2
Host Species Mouse
Immunogen The monoclonal antibody JC4 is specific for Cytotoxic necrotizing factor type 1 and the highly related Cytotoxic necrotizing factor type 2 (CNF1 and CNF2) of uropathogenic Escherichia coli.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.