missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNBP Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-983
This item is not returnable.
View return policy
Description
CNBP Polyclonal specifically detects CNBP in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CNBP | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CCHC-type zinc finger, nucleic acid binding protein, cellular nucleic acid binding protein, cellular nucleic acid-binding protein, DM2, erythroid differentiation-related, FLJ11631, PROMM, RNF163CNBP1ZNF9ZCCHC22, sterol regulatory element-binding protein, zinc finger protein 273, Zinc finger protein 9, zinc finger protein 9 (a cellular retroviral nucleic acid binding protein) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog . | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_003409.1 | |
| CNBP | |
| Peptide with sequence GESGHLARECTIE corresponding to C-Terminus according to NP_003409.1. | |
| 0.1 mg | |
| DNA replication Transcription Translation and Splicing | |
| 7555 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction