missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35835-100ul
This item is not returnable.
View return policy
Description
CNAP1 Polyclonal antibody specifically detects CNAP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CNAP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| CAPD2, CAP-D2, Chromosome condensation-related SMC-associated protein 1, Chromosome-associated protein D2, CNAP1XCAP-D2 homolog, hCAP-D2condensin complex subunit 1, KIAA0159chromosome condensation related SMC associated protein 1, Non-SMC condensin I complex subunit D2, non-SMC condensin I complex, subunit D2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1250-1350 of human CNAP1 (NP_055680.3).,, Sequence:, FGDKLSDESIFSAFLSVVGKLRRGAKPEGKAIIDEFEQKLRACHTRGLDGIKELEIGQAGSQRAPSAKKPSTGSRYQPLASTASDNDFVTPEPRRTTRRHP | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| 9918 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction