missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CN079 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559844
This item is not returnable.
View return policy
Description
Immunogen sequence: GGPWVTGTSA VPPSEPILSY ENILKCAFQE ITVQQAAEDV STIDHFLEIS SEEKPGVERV HKLCNESRKL Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.
C14orf79 gene ontology annotations related to this gene include intracellular transport.
Specifications
| CN079 | |
| Polyclonal | |
| Unconjugated | |
| CLBA1 | |
| C14orf79; chromosome 14 open reading frame 79; Clathrin-binding box of aftiphilin-containing protein 1; CLBA1; uncharacterized protein C14orf79; Uncharacterized protein CLBA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122616 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96F83 | |
| CLBA1 | |
| Recombinant protein corresponding to Human CN079. Recombinant protein control fragment (Product #RP-98463). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction