missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CMV pp65 peptide GPISGHVLK (HLA-A*1101)
Shop All IBA Lifesciences Products

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Specifications
Specifications
| Content And Storage | Recommended: -25°C to -15°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction