missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMTM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562589
This item is not returnable.
View return policy
Description
Immunogen sequence: MLSARDRRDR HPEEGVVAEL QGFAVDKAFL TSHKGI Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 64%.
This gene belongs to the chemokine-like factor gene superfamily. This family of genes encodes multi-pass membrane proteins that are similar to both the chemokine and the transmembrane 4 superfamilies of signaling molecules. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. Highly expressed in the brain.
Specifications
| CMTM5 | |
| Polyclonal | |
| Unconjugated | |
| CMTM5 | |
| 1500005P16Rik; 2900052H21Rik; chemokine-like factor super family 5; chemokine-like factor super family five variant 4; chemokine-like factor superfamily 5; chemokine-like factor superfamily member 5; CKLF like MARVEL transmembrane domain containing 5; CKLF-like MARVEL transmembrane domain containing 5; CKLF-like MARVEL transmembrane domain-containing protein 5; CKLFSF5; CMTM5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116173 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DZ9 | |
| CMTM5 | |
| Recombinant protein corresponding to Human CMTM5. Recombinant protein control fragment (Product #RP-99878). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction