missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMTM5, Mouse anti-Human, Clone: 2G1-6B10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00116173-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MLSARDRRDRHPEEGVVAELQGFAVDKAFLTSHKGILLETELALTLIIFICFTASISAYMAAALLEFFITLAFLFLYATQYYQRFDRINWPCLDFLRCVSAIIIFLVVSFAAVTSRDGAAIAAFVFGIILVSIFAYDAFKIYRTEMAPGASQGDQQSpecifications
| CMTM5 | |
| Monoclonal | |
| Unconjugated | |
| CKLF-like MARVEL transmembrane domain containing 5 | |
| CKLFSF5/FLJ37521 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2G1-6B10 | |
| In 1x PBS, pH 7.4 | |
| BC013109 | |
| CMTM5 | |
| CKLFSF5 (AAH13109, 1 a.a. ∼ 156 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 116173 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction