missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMTM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554914
This item is not returnable.
View return policy
Description
Immunogen sequence: GEELDGFEGE ASSTSMISGA SSPYQPTTEP VSQRRGLAGL RCDPDYL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].
Specifications
| CMTM4 | |
| Polyclonal | |
| Unconjugated | |
| CMTM4 | |
| chemokine-like factor super family 4; chemokine-like factor superfamily member 4; CKLF like MARVEL transmembrane domain containing 4; CKLF-like MARVEL transmembrane domain containing 4; CKLF-like MARVEL transmembrane domain-containing protein 4; Cklfsf4; Cmtm4; D19397; ENSMUSG00000051554; Gm9853 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZR5 | |
| CMTM4 | |
| Recombinant protein corresponding to Human CMTM4. Recombinant protein control fragment (Product #RP-91191). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction