Learn More
Invitrogen™ CMTM2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA568528
Product Code. 15826943
  • £440.00 / 100µL
Description
This target displays homology in the following species: Cow: 87%; Dog: 87%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 100%.
This gene belongs to the chemokine-like factor gene superfamily, a novel family that links the chemokine and the transmembrane 4 superfamilies of signaling molecules. The protein encoded by this gene may play an important role in testicular development.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CMTM2
Polyclonal
Unconjugated
CMTM2
1700001K04Rik; 1700041N15Rik; 1700063K20Rik; AI449770; ARR19; C32; chemokine-like factor super family 1; chemokine-like factor super family 2A; chemokine-like factor superfamily 2; chemokine-like factor superfamily 2-1b; chemokine-like factor superfamily member 2; Chemokine-like factor superfamily member 2A; Cklf; CKLF like MARVEL transmembrane domain containing 2; Cklf1; CKLF3; CKLF4; CKLF5; CKLF-like MARVEL transmembrane domain containing 2; CKLF-like MARVEL transmembrane domain containing 2A; CKLF-like MARVEL transmembrane domain-containing protein 2; CKLF-like MARVEL transmembrane domain-containing protein 2A; Cklfsf1; CKLFSF2; Cklfsf2-1b; Cklfsf2a; CMTM2; Cmtm2a; UCK-1; Unknown (protein for MGC:133644)
Rabbit
Affinity chromatography
RUO
146225
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8TAZ6
CMTM2
synthetic peptide directed towards the N terminal region of human CMTM2.