missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CMRL Medium, no glutamine
GREENER_CHOICE

Product Code. 11500576
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
500mL
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500576

Brand: Gibco™ 21530027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CMRL Medium, no glutamine

CMRL Medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L'cells under serum-free conditions. Gibco™ CMRL Medium is also especially useful for cloning monkey kidney cells and for growth of many other mammalian cell lines when supplemented with horse or calf serum.

Gibco™ CMRL Medium is a modified version of Medium 199. CMRL medium is particularly rich in nucleosides and vitamins.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

Dual-Site cGMP Manufacturing
Gibco™ CMRL Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ CMRL Medium product made in our Grand Island facility (11530-037). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

CMRL Medium contains no proteins or growth factors. Therefore, CMRL Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). CMRL Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type CMRL Medium
With Additives Phenol Red
Without Additives No Glutamine, No HEPES
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
Regulatory Status For in-vitro diagnostic use
Content And Storage 2 to 8°C
Description CMRL Medium, 500mL, Liquid Form, With Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.