missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMPK2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA559570
This item is not returnable.
View return policy
Description
Immunogen sequence: ECTSFIPEAR AVLDLVDQCP KQIQKGKFQV VAIEGLDATG KTTVTQSVAD SLKAVLLKSP PSCIGQWRKI FDDEPTIIRR AFYSLGNYIV ASEIAKESAK SPV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.
UMP-CMP kinase 2 (CMPK2) is the first nucleoside monophosphate kinase that has been identified in human mitochondria. It is a component of the salvage pathway for nucleotide synthesis that may participate in terminal differentiation of monocytic cells. There are two distinct domains of CMPK2: the N-terminal domain with unknown function and the C-terminal domain with the kinase function, suggesting that CMPK2 may be a bifunctional protein with other biological functions in addition to its UMP-CMP kinase activity. CMPK2 may participate in dUTP and dCTP synthesis and is responsible for phosphorylation of dCMP and dUMP in mitochondria. CMPK2 is actively involved in macrophage activation and the inflammatory response
Specifications
| CMPK2 | |
| Polyclonal | |
| Unconjugated | |
| CMPK2 | |
| TMPK2, TYKi, UMP-CMPK2, UMP-CMP kinase 2, mitochondrial, cytidylate kinase 2, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 129607 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5EBM0 | |
| CMPK2 | |
| Recombit protein corresponding to Human CMPK2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction