Learn More
CMPK1 Mouse anti-Human, CoraLite 594, Clone: 2D11G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67519-100UL
Description
Pyrimidine nucleoside monophosphate kinase [UMP/CMP kinase (UMP/CMPK)] (CMPK1) is also named as CMK, CMPK, UCK, UMK, UMPK and belongs to the adenylate kinase family. It plays a crucial role in the formation of UDP, CDP, and dCDP, which are required for cellular nucleic acid synthesis(PMID:11912132). CMPK1 can use a broad spectrum of phosphate donors but the best phosphate donors are ATP and dATP. CMPK1 has been reported as a prognostic marker for non-small cell lung cancer, pancreatic cancer and breast cancer (PMID: 21642870, 22838950, 27558661). The longest deduced protein contains 228 amino acids and has a calculated molecular mass of 26 kD. However, translation likely begins at the second initiation methionine, resulting in a protein of 196 amino acids(PMID:10462544). The shorter form is the endogenously translated protein(PMID:11912132). CMPK1 has 3 isoforms with the molecular mass of 22, 16 and 26 kDa.
Specifications
| CMPK1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Immunofluorescence | |
| 2D11G7 | |
| CoraLite 594 | |
| 100 μL | |
| 51727 | |
| Liquid |