missing translation for 'onlineSavingsMsg'
Learn More

CMPDA, MedChemExpress

Product Code. 16435228
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16435228 10 mg 10mg
16427947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16435228 Supplier Medchem Express Supplier No. HY12508
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CMPDA is a positive allosteric modulator of AMPA receptors with EC50s of 45.4 (plusmn) 4.2 nM/63.4 (plusmn) 5.6 nM for GluA2i/GluA2o receptor.

CMPDA is a positive allosteric modulator of AMPA receptors with EC50s of 45.4 (plusmn) 4.2 nM/63.4 (plusmn) 5.6 nM for GluA2i/GluA2o receptor.

Specifications

Chemical Name or Material CMPDA
CAS 380607-77-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H28N2O4S2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (265.58 mM)
SMILES O=S(NCCC1=CC=C(CCNS(=O)(C(C)C)=O)C=C1)(C(C)C)=O
Molecular Weight (g/mol) 376.53
Formula Weight 376.53
Percent Purity 97.19%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.