missing translation for 'onlineSavingsMsg'
Learn More

CMPDA, MedChemExpress

Product Code. 16427947
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16427947

Brand: Medchem Express HY12508

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CMPDA is a positive allosteric modulator of AMPA receptors with EC50s of 45.4 (plusmn) 4.2 nM/63.4 (plusmn) 5.6 nM for GluA2i/GluA2o receptor.

CMPDA is a positive allosteric modulator of AMPA receptors with EC50s of 45.4 (plusmn) 4.2 nM/63.4 (plusmn) 5.6 nM for GluA2i/GluA2o receptor.

Specifications

Chemical Name or Material CMPDA
CAS 380607-77-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H28N2O4S2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (265.58 mM)
SMILES O=S(NCCC1=CC=C(CCNS(=O)(C(C)C)=O)C=C1)(C(C)C)=O
Molecular Weight (g/mol) 376.53
Formula Weight 376.53
Percent Purity 97.19%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.