missing translation for 'onlineSavingsMsg'
Learn More

CMPD1, MedChemExpress

Product Code. 16466937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16466937 Supplier Medchem Express Supplier No. HY108643

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with apparent Ki (Kiapp) of 330 nM.

CMPD1 is a selective and non-ATP-competitive p38 MAPK-mediated MK2 phosphorylation inhibitor with apparent Ki (Kiapp) of 330 nM.

Specifications

Chemical Name or Material CMPD1
CAS 41179-33-3
Color Light Nude
Purity Grade Notes Research
Molecular Formula C22H20FNO2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (286.20 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(O)C=C1)CCCC2=CC=C(C3=CC=CC=C3F)C=C2
Molecular Weight (g/mol) 349.4
Formula Weight 349.4
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.