missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CMPD-1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CMPD-1 is a non-ATP-competitive, selective inhibitor of p38α-mediated MK2a (mitogen-activated protein kinase-2a) phosphorylation (apparent Ki = 330 nM). Does not inhibit p38α-mediated phosphorylation of MBP and ATF-2. Also tubulin polymerization inhibitor. Cytotoxic in glioblastoma cells at concentrations that do not affect the MK2 pathway.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | CMPD-1 |
| Purity | >99% |
| Molecular Weight (g/mol) | 349.4 |
| Inhibitors | CMPD-1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction