missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CMPD-1
GREENER_CHOICE

Product Code. 18084522
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18084522

Brand: Tocris Bioscience™ 2186/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of p38α-mediated MK2a phosphorylation; also tubulin polymerization inhibitor

CMPD-1 is a non-ATP-competitive, selective inhibitor of p38α-mediated MK2a (mitogen-activated protein kinase-2a) phosphorylation (apparent Ki = 330 nM). Does not inhibit p38α-mediated phosphorylation of MBP and ATF-2. Also tubulin polymerization inhibitor. Cytotoxic in glioblastoma cells at concentrations that do not affect the MK2 pathway.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type CMPD-1
Purity >99%
Molecular Weight (g/mol) 349.4
Inhibitors CMPD-1

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.