missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CMP-Azido-Sialic Acid
Description
Includes
Incorporate or detect sialic acid without expensive specialized equipment!
Applications:
- For in vitro enzymatic incorporation of azido-sugars into specific, targeted glycans.
- Detecting the presence or absence of sialated glyca
Provided in 20mM Tris, pH 8.0
Specifications
Specifications
| Quantity | 100 μg |
| Molecular Formula | C20H30N7O15P |
| Formula Weight | 639.46 Da |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?