missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMKLR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595744
This item is not returnable.
View return policy
Description
Immunogen sequence: NRLAKTKKPF KIIVTIIITF FLCWCPYHTL NLLELHHTAM PGSVFSLGLP LATALAIANS CMNPILYVFM GQDFKKFKVA LFSRLVNALS EDTGHSSYPS H; Positive Samples: Rat heart; Cellular Location: Cell membrane, Multi-pass membrane protein.
Chemokine-like receptor 1 (CMKLR1) is a receptor for the chemoattractant adipokine chemerin/RARRES2 and for the omega-3 fatty acid derived molecule resolvin E1. Interaction with RARRES2 induces activation of intracellular signaling molecules, such as SKY, MAPK1/3 (ERK1/2), MAPK14/P38MAPK and PI3K leading to multifunctional effects, like, reduction of immune responses, enhancing of adipogenesis and angionesis. Resolvin E1 down-regulates cytokine production in macrophages by reducing the activation of MAPK1/3 (ERK1/2) and NF-kappa-B. CMKLR1 positively regulates adipogenesis and adipocyte metabolism and acts as a coreceptor for several SIV strains (SIVMAC316, SIVMAC239, SIVMACL7E-FR and SIVSM62A), as well as a primary HIV-1 strain (92UG024-2).
Specifications
| CMKLR1 | |
| Polyclonal | |
| Unconjugated | |
| CMKLR1 | |
| chemerin chemokine-like receptor 1; chemerin receptor; CHEMERINR; chemokine receptor-like 1; chemokine-like receptor 1; CHEMR23; CMKLR1; Dez; Gpcr27; G-protein coupled chemoattractant-like receptor; G-protein coupled receptor 27; G-protein coupled receptor ChemR23; G-protein coupled receptor DEZ; mcmklr1; orphan G-protein coupled receptor, Dez; resolvin E1 receptor; RVER1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1240, 14747, 60669 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.16 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O35786, P97468, Q99788 | |
| CMKLR1 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human CMKLR1 (NP_0011358151). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction