missing translation for 'onlineSavingsMsg'
Learn More

CMK, MedChemExpress

Product Code. 16480608
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480608 5 mg 5mg
16458158 10 mg 10mg
16450356 2 mg 2mg
16403766 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480608 Supplier Medchem Express Supplier No. HY52101
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CMK is a RSK2 kinase inhibitor which exhibits similar potency but less chemical stability compared with FMK.

CMK is a RSK2 kinase inhibitor which exhibits similar potency but less chemical stability compared with FMK.

Specifications

Chemical Name or Material CMK
CAS 821794-90-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H19ClN4O2
Quantity 5 mg
Solubility Information DMSO : 150 mg/mL (418.04 mM; Need ultrasonic)
SMILES NC1=C(C2=NC=N1)C(C3=CC=C(C=C3)C)=C(N2CCCO)C(CCl)=O
Molecular Weight (g/mol) 358.82
Formula Weight 358.82
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.