missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMG2 Monoclonal Antibody (406510)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561227
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Anthrax toxin receptor 2 (ATR2), also designated capillary morphogenesis gene 2 (CMG2), is a cellular receptor for anthrax toxin. ATR2/CMG2 is a type 1 membrane protein that includes an extracellular von Willebrand factor A (VWA) domain with a metal ion-dependent adhesion site (MIDAS) motif. ATR2/CMG2 binds to protective antigen (PA) which is one of the three monomeric proteins which are released as anthrax toxin from Bacillus anthracis. Once bound, PA facilitates the delivery of the other two proteins, edema factor (EF) and lethal factor (LF), into the cytosol. Mutations in the gene ATR2/CMG2 result in inhibition of binding by the vWA domain, associated with infantile systemic hyalinosis (ISH) and juvenile hyaline fibromatosis (JHF). ISH and JHF are rare stiff-skin syndromes characterized by hyaline desposition in various organs, especially the skin and gingiva.
Specifications
| CMG2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P58335 | |
| ANTXR2 | |
| Mouse myeloma cell line NS0-derived recombinant human CMG-2/ANTXR2, Ser22-Arg200. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 406510 | |
| Unconjugated | |
| ANTXR2 | |
| 2310046B19Rik; anthrax toxin receptor 2; ANTXR cell adhesion molecule 2; ANTXR cell adhesion molecule 2; anthrax toxin receptor 2; ANTXR2; AW561899; Capillary morphogenesis gene 2 protein; capillary morphogenesis protein 2; cI-35; CMG2; CMG-2; FLJ31074; HFS; ISH; JHF; MGC111533; MGC45856 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 118429 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction