missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMG-2/ANTXR2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49866
This item is not returnable.
View return policy
Description
CMG-2/ANTXR2 Polyclonal specifically detects CMG-2/ANTXR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| CMG-2/ANTXR2 | |
| Polyclonal | |
| Immunohistochemistry 2.5 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/mL, Peptide ELISA Detection limit 1:16000 | |
| P58335 | |
| ANTXR2 | |
| Peptide with sequence C-HEGLKLANEQIQK corresponding to internal region according to NP_477520.2, NP_001139266.1. | |
| 0.1 mg | |
| Neuroscience | |
| 118429 | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| anthrax toxin receptor 2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_477520.2; NP_001139266.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction