missing translation for 'onlineSavingsMsg'
Learn More

CMG-2/ANTXR2 Antibody (406510) [FITC], Novus Biologicals™

Product Code. 30734990 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734990 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734990 Supplier Novus Biologicals Supplier No. FAB29401F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CMG-2/ANTXR2 Monoclonal antibody specifically detects CMG-2/ANTXR2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMG-2/ANTXR2
Applications ELISA
Classification Monoclonal
Clone 406510
Conjugate FITC
Formulation PBS
Gene Alias anthrax toxin receptor 2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CMG-2/ANTXR2, Ser22-Arg200, Accession # P58335
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 118429
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.