missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24030-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| CMC1 | |
| Polyclonal | |
| Unconjugated | |
| CMC1 | |
| C3orf68, CMC1, Cmc1p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 152100, 67899 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.69 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z7K0, Q9CPZ8 | |
| Cmc1 | |
| CMC1 Fusion Protein Ag21224 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction