missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMBL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557724
This item is not returnable.
View return policy
Description
Immunogen sequence: AYPCPCDIGH RLEYGGLGRE VQVEHIKAYV TKSPVDAGKA VIVIQDIFGW QLPNTRYIAD MISGNGYTTI VPDFFVG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
CMBL is a cysteine hydrolase of the dienelactone hydrolase family that is highly expressed in liver cytosol. CMBL preferentially cleaves cyclic esters, and it activates medoxomil-ester prodrugs in which the medoxomil moiety is linked to an oxygen atom.
Specifications
| CMBL | |
| Polyclonal | |
| Unconjugated | |
| Cmbl | |
| 2310016A09Rik; Ab2-225; carboxymethylenebutenolidase homolog; carboxymethylenebutenolidase homolog (Pseudomonas); carboxymethylenebutenolidase-like (Pseudomonas); CMBL; JS-1; Liver regeneration-related protein LRRG072; RGD1306952 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 134147 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DG6 | |
| Cmbl | |
| Recombinant protein corresponding to Human CMBL. Recombinant protein control fragment (Product #RP-96951). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction