missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CMBL Polyclonal antibody specifically detects CMBL in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | CMBL |
| Classification | Polyclonal |
| Purification Method | Antigen and protein A Affinity-purified |
| Research Discipline | metabolism, Proteases & Other Enzymes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?