missing translation for 'onlineSavingsMsg'
Learn More

CMBL Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18428879 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18428879 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428879 Supplier Novus Biologicals Supplier No. NBP299788JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CMBL Polyclonal antibody specifically detects CMBL in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Janelia Fluor 646
Research Discipline metabolism, Proteases & Other Enzymes
Target Species Human
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.