missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMBL Antibody (2B11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22618DL594
This item is not returnable.
View return policy
Description
CMBL Monoclonal antibody specifically detects CMBL in Human samples. It is validated for Western Blot, ELISA
Specifications
| CMBL | |
| Monoclonal | |
| DyLight 594 | |
| carboxymethylenebutenolidase homolog, carboxymethylenebutenolidase homolog (Pseudomonas), carboxymethylenebutenolidase-like (Pseudomonas), EC 3.1, FLJ23617 | |
| Recombinant human CMBL (1-245aa) purified from E. coli | |
| 0.1 mL | |
| metabolism, Proteases & Other Enzymes | |
| 134147 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 2B11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction