missing translation for 'onlineSavingsMsg'
Learn More

CMAS Antibody (2E1), Novus Biologicals™

Product Code. 18398809 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398809 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398809 Supplier Novus Biologicals Supplier No. H00055907M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CMAS Monoclonal antibody specifically detects CMAS in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMAS
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2E1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061156
Gene Alias CMP-N-acetylneuraminic acid synthase, CMP-N-acetylneuraminic acid synthetase, CMP-Neu5Ac synthetase, CMP-NeuNAc synthase, CMP-NeuNAc synthetase, cytidine 5'-monophosphate N-acetylneuraminic acid synthetase, cytidine monophosphate N-acetylneuraminic acid synthetase, EC 2.7.7.43, N-acylneuraminate cytidylyltransferase
Host Species Mouse
Immunogen CMAS (NP_061156, 164 a.a. ~ 263 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EGYDSVFSVVRRHQFRWSEIQKGVREVTEPLNLNPAKRPRRQDWDGELYENGSFYFAKRHLIEMGYLQGGKMAYYEMRAEHSVDIDVDIDWPIAEQRVLR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55907
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.