missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CMA
GREENER_CHOICE

Product Code. 18795386
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18795386

Brand: Tocris Bioscience™ 6677/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Murine-selective STING agonist

CMA is a STING agonist; murine-selective (Kd = 3.5 μM). Triggers type I IFN response in murine macrophages, with no effect on human STING signaling.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Oxo-10(9H)-acridineacetic acid
CAS 38609-97-1
Synonym ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
Quantity 50 mg
Target STING Agonists
Molecular Formula C15H11NO3
Formula Weight 253.25
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.