missing translation for 'onlineSavingsMsg'
Learn More

CM-579 trihydrochloride, MedChemExpress

Product Code. 16451137
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451137 50 mg 50mg
16427637 5 mg 5mg
16425666 100 mg 100mg
16414888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451137 Supplier Medchem Express Supplier No. HY117421A
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CM-579 trihydrochloride is a first-in-class reversible, dual inhibitor of G9a and DNMT, with IC50 values of 16 nM, 32 nM for G9a and DNMT, respectively. Has potent in vitro cellular activity in a wide range of cancer cells.

CM-579 trihydrochloride is a first-in-class reversible, dual inhibitor of G9a and DNMT, with IC50 values of 16 nM, 32 nM for G9a and DNMT, respectively. Has potent in vitro cellular activity in a wide range of cancer cells.

Specifications

Chemical Name or Material CM-579 trihydrochloride
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C29H43Cl3N4O3
Quantity 50 mg
Solubility Information H2O : 37.5 mg/mL (62.29 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (55.36 mM; Need ultrasonic)
SMILES CN1CCC(CNC2=CC(C3=CC=C(C)O3)=NC4=CC(OCCCN5CCCC5)=C(OC)C=C24)CC1.Cl.Cl.Cl
Molecular Weight (g/mol) 602.04
Formula Weight 602.04
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.