missing translation for 'onlineSavingsMsg'
Learn More

CM-579 trihydrochloride, MedChemExpress

Product Code. 16414888
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414888

Brand: Medchem Express HY117421A10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CM-579 trihydrochloride is a first-in-class reversible, dual inhibitor of G9a and DNMT, with IC50 values of 16 nM, 32 nM for G9a and DNMT, respectively. Has potent in vitro cellular activity in a wide range of cancer cells.

CM-579 trihydrochloride is a first-in-class reversible, dual inhibitor of G9a and DNMT, with IC50 values of 16 nM, 32 nM for G9a and DNMT, respectively. Has potent in vitro cellular activity in a wide range of cancer cells.

Specifications

Chemical Name or Material CM-579 trihydrochloride
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C29H43Cl3N4O3
Quantity 10 mg
Solubility Information H2O : 37.5 mg/mL (62.29 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (55.36 mM; Need ultrasonic)
SMILES CN1CCC(CNC2=CC(C3=CC=C(C)O3)=NC4=CC(OCCCN5CCCC5)=C(OC)C=C24)CC1.Cl.Cl.Cl
Molecular Weight (g/mol) 602.04
Formula Weight 602.04
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.