missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Clusterin (CLU) ELISA Kit

Product Code. 17135909
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 17135909

Brand: Invitrogen™ ERCLUX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Rat Clusterin (Rt CLU) ELISA quantitates Rt Clusterin in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt Clusterin. Principle of the method The Rat Clusterin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Isoform 1 functions as extracellular chaperone that prevents aggregation of nonnative proteins. Prevents stress-induced aggregation of blood plasma proteins. Inhibits formation of amyloid fibrils by APP, APOC2, B2M, CALCA, CSN3, SNCA and aggregation-prone LYZ variants (in vitro). Does not require ATP. Maintains partially unfolded proteins in a state appropriate for subsequent refolding by other chaperones, such as HSPA8/HSC70. Does not refold proteins by itself. Binding to cell surface receptors triggers internalization of the chaperone-client complex and subsequent lysosomal or proteasomal degradation. Secreted isoform 1 protects cells against apoptosis and against cytolysis by complement. Intracellular isoforms interact with ubiquitin and SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complexes and promote the ubiquitination and subsequent proteasomal degradation of target proteins. Promotes proteasomal degradation of COMMD1 and IKBKB. Modulates NF-kappa-B transcriptional activity. Nuclear isoforms promote apoptosis. Mitochondrial isoforms suppress BAX-dependent release of cytochrome c into the cytoplasm and inhibit apoptosis. Plays a role in the regulation of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05371
Assay Range 1.311 to 320 ng/mL
Assay Sensitivity 1.3 ng/mL
Calibration Range 1.3 ng/ml to 320 ng/ml
Description Clusterin (CLU) ELISA Kit, 96 Assays, 1 Kit, Rat Species, Histones Protein Family, Oncology Research Discipline, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of Clusterin (Clu) in Samples, Using a Microplate Reader
Detectable Analytes Clusterin (CLU)
Assay Solid-phase sandwich ELISA
Conjugate HRP
Detection Method Colorimetric
Product Type ELISA
Protein Family Histones
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects rat and mouse Clusterin. Other species not determined.
Substrate Type TMB
Sufficient For 96 assays
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 24854
Gene Symbol Clu
Host Species Rat
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Oncology
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym APOJ, CLI, DAG, RATTRPM2B, SGP-2, SGP2, SP-40, SP40, TRPM-2, TRPM2B, Trpm2, Trpmb
Target Apolipoprotein J
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.