missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clusterin/APOJ Rabbit anti-Human, Clone: 13, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89884-50UL
This item is not returnable.
View return policy
Description
Clusterin/APOJ Monoclonal specifically detects Clusterin/APOJ in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Clusterin/APOJ | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 13 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| 40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2 | |
| Recombinant Human Clusterin/APOJ protein | |
| 50 μg | |
| Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity | |
| 1191 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction