missing translation for 'onlineSavingsMsg'
Learn More

CLUH Rabbit, anti-Human, Polyclonal, Proteintech

Product Code. 18056175 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18056175 150 μL 150µL
18086375 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18056175 Supplier Proteintech Supplier No. 309861AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CLUH antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CLUH. This antibody recognizes human, mouse antigen. The CLUH antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen CLUH
Applications Western Blot, ELISA
Concentration 150 μg/mL
Gene Symbols CLU1, DKFZp686L05242, DKFZp686M09233, FLJ23672, KIAA0664, Protein KIAA0664
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 23277
Target Species Human, Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.