missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to CLTCL1. It has no cross reaction to CLTC.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
Specifications
| Antigen | CLTCL1 |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CLTCL1 |
| Gene Accession No. | P53675 |
| Gene Alias | CHC22, Clathrin heavy chain 2, clathrin, heavy chain like 1, CLH 22, CLH22, CLTCL, CLTCL1, CLTD |
| Gene Symbols | CLTCL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?