Learn More
Invitrogen™ CLTCL1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA568407
Product Code. 15833234
  • £441.00 / 100µL
Description
CLTCL1 Polyclonal Antibody for IHC (P)
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLTCL1
Polyclonal
Unconjugated
CLTCL1
CHC22; clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 22; clathrin, heavy chain-like 1; Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; CLH22; CLH-22; CLTCL; CLTCL1; CLTD
Rabbit
Antigen affinity chromatography
RUO
8218
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P53675
CLTCL1
Synthetic peptide corresponding to residues near the C terminal of human CLTCL1.