missing translation for 'onlineSavingsMsg'
Learn More

CLTC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1B3D7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66487-1-IG-20UL

 View more versions of this product

Product Code. 16888405

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.
Specifications

Specifications

CLTC
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11442, Q00610, Q68FD5
Cltc
CLTC Fusion Protein Ag25040
20 μL
Primary
Human, Mouse, Rat, Pig
Antibody
IgG1
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1B3D7
Unconjugated
CLTC
CHC, CHC17, Clathrin heavy chain 1, clathrin, heavy chain (Hc), CLH 17, CLH17, CLTC, CLTCL2, Hc, KIAA0034
Mouse
Protein G
RUO
100271929, 1213, 54241, 67300
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.