missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1B3D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66487-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.Specifications
| CLTC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11442, Q00610, Q68FD5 | |
| Cltc | |
| CLTC Fusion Protein Ag25040 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1B3D7 | |
| Unconjugated | |
| CLTC | |
| CHC, CHC17, Clathrin heavy chain 1, clathrin, heavy chain (Hc), CLH 17, CLH17, CLTC, CLTCL2, Hc, KIAA0034 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100271929, 1213, 54241, 67300 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction