Learn More
Invitrogen™ CLSTN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590686
Description
Immunogen sequence: SLQQRGLELT NTSAYLTIAG VESITVYEEI LRQARYRLRH GAALYTRKFR LSCSEMNGRY SSNEFIVEVN VLHSMNRVAH PSHVLSSQQF LHRGHQPPPE MAGHSLASSH RNSMIPSAAT L; Positive Samples: Mouse brain; Cellular Location: Cell membrane, Endoplasmic reticulum membrane, Golgi apparatus membrane, Single-pass type I membrane protein.
Specifications
| CLSTN3 | |
| Polyclonal | |
| Unconjugated | |
| Clstn3 | |
| alcadein beta; alcadein-beta; alcbeta; Alc-beta; cadherin-related family member 14; calsyntenin 3; calsyntenin-3; CDHR14; CLSTN3; Cs3; Cst-3; Cstn3; KIAA0726; KIAA0726 MGC131797; MGC138488; mKIAA0726 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232370 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.99 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99JH7 | |
| Clstn3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 730-850 of human CLSTN3 (NP_0555332). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.