missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPX Antibody (S05-2A6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19849-50ul
This item is not returnable.
View return policy
Description
CLPX Monoclonal antibody specifically detects CLPX in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CLPX | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S05-2A6 | |
| Unconjugated | |
| ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial, ClpX caseinolytic peptidase X homolog (E. coli), ClpX caseinolytic protease X homolog, energy-dependent regulator of proteolysis | |
| A synthetic peptide of human CLPX (Uniprot # O76031) | |
| 50 μg | |
| Stem Cell Markers | |
| 10845 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction