missing translation for 'onlineSavingsMsg'
Learn More

CLPX Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£161.00 - £383.00

Specifications

Antigen CLPX
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30227887
View Documents
Novus Biologicals
NBP3-33213-100ul
100 μL
£383.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30226806
View Documents
Novus Biologicals
NBP3-33213-20ul
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CLPX Monoclonal antibody specifically detects CLPX in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

CLPX
ELISA, Western Blot
Unconjugated
Rabbit
Stem Cell Markers
PBS (pH 7.3), 50% glycerol, 0.05% BSA
10845
IgG
Affinity purified
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial, ClpX caseinolytic peptidase X homolog (E. coli), ClpX caseinolytic protease X homolog, energy-dependent regulator of proteolysis
A synthetic peptide corresponding to a sequence within amino acids 534-633 of human CLPX (O76031).,, Sequence:, SMDKCELNVTEDALKAIARLALERKTGARGLRSIMEKLLLEPMFEVPNSDIVCVEVDKEVVEGKKEPGYIRAPTKESSEEEYDSGVEEEGWPRQADAANS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.