missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLPTM1 Recombinant Rabbit Monoclonal Antibody (K01_1P65)

Product Code. 30825938
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825938 50 μL
30830128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825938 Supplier Invitrogen™ Supplier No. MA565067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CLPTM1 Recombinant Monoclonal Antibody for Western Blot

CLPTM1 may play a role in T-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPTM1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1P65
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene CLPTM1
Gene Accession No. O96005
Gene Alias cleft lip and palate associated transmembrane protein 1; cleft lip and palate transmembrane protein 1; Cleft lip and palate transmembrane protein 1 homolog; CLPTM1; CLPTM1, transmembrane protein; HS9; N14; Thymic epithelial cell surface antigen
Gene Symbols CLPTM1
Host Species Rabbit
Immunogen A synthetic peptide of human CLPTM1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1209
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.