missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLPTM1 Recombinant Rabbit Monoclonal Antibody (23GB3090), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553321
This item is not returnable.
View return policy
Description
CLPTM1 may play a role in T-cell development.
Specifications
| CLPTM1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O96005, Q8VBZ3 | |
| CLPTM1 | |
| A synthesized peptide derived from CLPTM1 (30-54AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3090 | |
| Unconjugated | |
| CLPTM1 | |
| cleft lip and palate associated transmembrane protein 1; cleft lip and palate transmembrane protein 1; Cleft lip and palate transmembrane protein 1 homolog; CLPTM1; CLPTM1, transmembrane protein; HS9; N14; Thymic epithelial cell surface antigen | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1209, 292696, 56457 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction