missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPTM1 Antibody (SR2274), Novus Biologicals™
Rabbit Monoclonal Antibody
£272.00 - £351.00
Specifications
| Antigen | CLPTM1 |
|---|---|
| Clone | SR2274 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666454
|
Novus Biologicals
NBP3-22486-100ul |
100 μg |
£351.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619254
|
Novus Biologicals
NBP3-22486-50ul |
50 μg |
£272.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLPTM1 Monoclonal antibody specifically detects CLPTM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| CLPTM1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cleft lip and palate associated transmembrane protein 1, cleft lip and palate transmembrane protein 1 | |
| A synthesized peptide derived from human CLPTM1 (Uniprot #: O96005) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2274 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1209 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title